Assay Details
Target Gene Details
Entrez Gene ID: | 26151 |
Gene Name: | N-acetyltransferase 9 (putative) |
Gene Aliases: |
EBSP, hNATL |
Location: |
Chr.17:74770529-74776425 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT9 | NM_001305077.1 | 1 | 100 | NP_001292006.1 |
| NM_001305078.1 | 1 | 100 | NP_001292007.1 | |
| NM_001305079.1 | 1 | 100 | NP_001292008.1 | |
| NM_001305080.1 | 1 | 100 | NP_001292009.1 | |
| NM_001305081.1 | 1 | 100 | NP_001292010.1 | |
| NM_001305082.1 | 1 | 100 | NP_001292011.1 | |
| NM_001305083.1 | 1 | 100 | NP_001292012.1 | |
| NM_001305084.1 | 1 | 100 | NP_001292013.1 | |
| NM_001305085.1 | 1 | 100 | NP_001292014.1 | |
| NM_001305086.1 | 1 | 100 | NP_001292015.1 | |
| NM_001305087.1 | 1 | 100 | NP_001292016.1 | |
| NM_001305088.1 | 1 | 100 | NP_001292017.1 | |
| NM_015654.4 | 1 | 100 | NP_056469.2 | |
| NR_130953.1 | 1 | 100 | ||
| NR_130954.1 | 1 | 100 | ||
| XM_005257212.4 | XP_005257269.1 | |||
| XM_011524607.2 | XP_011522909.1 | |||
| XM_017024437.1 | XP_016879926.1 | |||
| XM_017024438.1 | XP_016879927.1 | |||
| XM_017024439.1 | XP_016879928.1 | |||
| AK123115.1 | ||||
| AK225602.1 | ||||
| AK308569.1 | ||||
| AK312958.1 | ||||
| AL050269.1 | CAB43370.1 | |||
| AL537925.3 | ||||
| BC004195.2 | AAH04195.1 | |||
| BC004225.2 | AAH04225.1 | |||
| BG702300.1 | ||||
| BI821280.1 | ||||
| BM464121.1 | ||||
| BM909550.1 | ||||
| BP197633.1 | ||||
| BP231776.1 | ||||
| BQ068039.1 | ||||
| BU159218.1 | ||||
| BX333236.2 | ||||
| DA010490.1 | ||||
| DA259097.1 | 1 | 100 | ||
| DB171198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map