Assay Details
Target Gene Details
| Entrez Gene ID: | 7869 | 
| Gene Name: | semaphorin 3B | 
| Gene Aliases: | LUCA-1, SEMA5, SEMAA, SemA, semaV | 
| Location: | Chr.3:50267558-50277171 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| SEMA3B | NM_001005914.2 | 1 | 143 | NP_001005914.1 | 
| NM_004636.3 | 1 | 143 | NP_004627.1 | |
| AK308875.1 | 1 | 146 | ||
| BI603095.1 | 1 | 136 | ||
| HY146744.1 | 1 | 143 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2759148 | Chr.3:50161045 - 50333946 on Build GRCh38 | Loss |  TUSC2  IFRD2  GNAT1  SEMA3B-AS1  RASSF1  SEMA3F  GNAI2  MIR6872  HYAL2  SEMA3B  MIR566  LSMEM2  SLC38A3  MIR5787  HYAL1  NAT6  HYAL3 | 
| nsv523560 | Chr.3:50134964 - 50314769 on Build GRCh38 | Loss |  IFRD2  GNAT1  SEMA3B-AS1  SEMA3F-AS1  SEMA3F  GNAI2  MIR6872  SEMA3B  MIR566  LSMEM2  SLC38A3  LOC107986035  MIR5787  HYAL1  NAT6  HYAL3 | 
| dgv602n27 | Chr.3:50140578 - 50287241 on Build GRCh38 | Loss |  MIR566  LSMEM2  SLC38A3  GNAT1  LOC107986035  MIR5787  SEMA3B-AS1  SEMA3F-AS1  SEMA3F  GNAI2  MIR6872  SEMA3B | 
| dgv8317n54 | Chr.3:50206077 - 50383123 on Build GRCh38 | Loss |  NPRL2  TUSC2  IFRD2  SEMA3B-AS1  RASSF1  GNAI2  MIR6872  HYAL2  SEMA3B  CYB561D2  LSMEM2  SLC38A3  MIR5787  CACNA2D2  HYAL1  NAT6  HYAL3  ZMYND10  TMEM115  RASSF1-AS1 | 
| nsv834693 | Chr.3:50251400 - 50470533 on Build GRCh38 | Loss |  NPRL2  TUSC2  IFRD2  SEMA3B-AS1  RASSF1  GNAI2  MIR6872  HYAL2  SEMA3B  CYB561D2  LSMEM2  CACNA2D2  HYAL1  NAT6  HYAL3  ZMYND10  TMEM115  RASSF1-AS1 | 
| nsv834692 | Chr.3:50161052 - 50356033 on Build GRCh38 | Loss |  NPRL2  TUSC2  IFRD2  GNAT1  SEMA3B-AS1  RASSF1  SEMA3F  GNAI2  MIR6872  HYAL2  SEMA3B  CYB561D2  MIR566  LSMEM2  SLC38A3  MIR5787  HYAL1  NAT6  HYAL3  ZMYND10  TMEM115  RASSF1-AS1 | 
| dgv8316n54 | Chr.3:50115923 - 50299991 on Build GRCh38 | Loss |  RBM5  IFRD2  GNAT1  SEMA3B-AS1  SEMA3F-AS1  SEMA3F  GNAI2  MIR6872  SEMA3B  MIR566  LSMEM2  SLC38A3  LOC107986035  MIR5787  HYAL1  NAT6  HYAL3 | 
| nsv470561 | Chr.3:50121341 - 50547568 on Build GRCh38 | Loss |  SEMA3B-AS1  SEMA3F-AS1  MIR6872  CYB561D2  MIR566  SLC38A3  LOC107986035  HYAL3  RASSF1-AS1  NPRL2  TUSC2  IFRD2  GNAT1  RASSF1  SEMA3F  GNAI2  HYAL2  SEMA3B  LSMEM2  MIR5787  CACNA2D2  HYAL1  NAT6  ZMYND10  TMEM115 | 
| nsv523662 | Chr.3:50107518 - 50547568 on Build GRCh38 | Loss |  RBM5  SEMA3B-AS1  SEMA3F-AS1  MIR6872  CYB561D2  MIR566  SLC38A3  LOC107986035  HYAL3  RASSF1-AS1  NPRL2  TUSC2  IFRD2  GNAT1  RASSF1  SEMA3F  GNAI2  HYAL2  SEMA3B  LSMEM2  MIR5787  CACNA2D2  HYAL1  NAT6  ZMYND10  TMEM115 | 
| nsv590255 | Chr.3:50210391 - 50399396 on Build GRCh38 | Loss |  NPRL2  TUSC2  IFRD2  SEMA3B-AS1  RASSF1  GNAI2  MIR6872  HYAL2  SEMA3B  CYB561D2  LSMEM2  SLC38A3  MIR5787  CACNA2D2  HYAL1  NAT6  HYAL3  ZMYND10  TMEM115  RASSF1-AS1 | 
| nsv954855 | Chr.3:50152468 - 50439169 on Build GRCh38 | Deletion |  SEMA3B-AS1  SEMA3F-AS1  MIR6872  CYB561D2  MIR566  SLC38A3  LOC107986035  HYAL3  RASSF1-AS1  NPRL2  TUSC2  IFRD2  GNAT1  RASSF1  SEMA3F  GNAI2  HYAL2  SEMA3B  LSMEM2  MIR5787  CACNA2D2  HYAL1  NAT6  ZMYND10  TMEM115 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top