Assay Details
Target Gene Details
Entrez Gene ID: | 5564 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit beta 1 |
Gene Aliases: |
AMPK, HAMPKb |
Location: |
Chr.12:119667864-119681624 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAB1 | NM_006253.4 | 7 | 1796 | NP_006244.2 |
| XM_005253909.1 | 8 | 1698 | XP_005253966.1 | |
| AF022116.1 | 7 | 1774 | AAC98897.1 | |
| AK127820.1 | 2 | 1850 | ||
| BC001007.2 | 7 | 1773 | AAH01007.1 | |
| BC001056.2 | 7 | 1773 | AAH01056.1 | |
| BC001823.2 | 7 | 1773 | AAH01823.1 | |
| BC017671.1 | 7 | 1782 | AAH17671.1 | |
| BX537486.1 | 2 | 3419 | ||
| U83994.1 | 7 | 1752 | AAD09237.1 |
More Information
Set Membership: |
|

Genomic Map