Assay Details
Target Gene Details
Entrez Gene ID: | 55139 |
Gene Name: | ankyrin repeat and zinc finger domain containing 1 |
Gene Aliases: |
ZNF744 |
Location: |
Chr.2:219229757-219236669 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKZF1 | NM_001042410.1 | NP_001035869.1 | ||
| NM_001282792.1 | NP_001269721.1 | |||
| NM_018089.2 | NP_060559.2 | |||
| XM_005246663.2 | XP_005246720.1 | |||
| XM_011511392.2 | XP_011509694.1 | |||
| XM_017004420.1 | XP_016859909.1 | |||
| XM_017004421.1 | XP_016859910.1 | |||
| AF364318.1 | ||||
| AK001277.1 | BAA91596.1 | |||
| AK023206.1 | BAB14462.1 | |||
| AK095304.1 | ||||
| AK098282.1 | ||||
| AK225275.1 | ||||
| AK225309.1 | ||||
| AK303077.1 | ||||
| AK303537.1 | ||||
| BC000238.1 | AAH00238.1 | |||
| BC008948.2 | AAH08948.1 | |||
| DQ412564.1 | ABD65410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834543 | Chr.2:219226398 - 219416430 on Build GRCh38 | Loss |
|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
| nsv508885 | Chr.2:219231225 - 219333602 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map