Assay Details
Target Gene Details
Entrez Gene ID: | 10622 |
Gene Name: | RNA polymerase III subunit G |
Gene Aliases: |
RPC32, RPC7 |
Location: |
Chr.5:90473929-90514557 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3G | NM_006467.2 | NP_006458.2 | ||
| XM_005248404.3 | XP_005248461.1 | |||
| XM_011543100.2 | XP_011541402.1 | |||
| XM_011543101.2 | XP_011541403.1 | |||
| XM_011543102.2 | XP_011541404.1 | |||
| XM_011543103.2 | XP_011541405.1 | |||
| XM_011543104.2 | XP_011541406.1 | |||
| XM_017008957.1 | XP_016864446.1 | |||
| AK295434.1 | ||||
| BM788728.1 | ||||
| BP348952.1 | ||||
| U93868.1 | AAB63676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518131 | Chr.5:90452325 - 90485777 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map