Assay Details
Target Gene Details
Entrez Gene ID: | 92736 |
Gene Name: | otopetrin 2 |
Gene Aliases: |
- |
Location: |
Chr.17:74922963-74933913 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OTOP2 | XM_011525479.1 | XP_011523781.1 |
Target Gene Details
Entrez Gene ID: | 124590 |
Gene Name: | USH1 protein network component sans |
Gene Aliases: |
ANKS4A, SANS |
Location: |
Chr.17:74916083-74923263 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USH1G | NM_001282489.2 | 1 | 175 | NP_001269418.1 |
NM_173477.4 | 1 | 175 | NP_775748.2 | |
AK091243.1 | 1 | 168 | BAC03619.1 | |
AK289804.1 | 1 | 172 | ||
AK296899.1 | 1 | 168 | ||
BC101096.2 | AAI01097.1 | |||
BC101097.1 | AAI01098.1 | |||
BC101098.2 | AAI01099.1 | |||
BC101099.1 | AAI01100.1 | |||
DC403891.1 | 1 | 175 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576026 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1056788 | Chr.17:74922211 - 75028689 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
