Assay Details
Target Gene Details
Entrez Gene ID: | 8613 |
Gene Name: | phospholipid phosphatase 3 |
Gene Aliases: |
Dri42, LPP3, PAP2B, PPAP2B, VCIP |
Location: |
Chr.1:56494747-56579584 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP3 | NM_003713.4 | 6 | 1882 | NP_003704.3 |
| AF480883.1 | 7 | 1694 | AAO84481.1 | |
| BM719257.1 | ||||
| BU608248.1 | 1 | 367 |
More Information
Set Membership: |
|

Genomic Map