Assay Details
Target Gene Details
Entrez Gene ID: | 10518 |
Gene Name: | calcium and integrin binding family member 2 |
Gene Aliases: |
DFNB48, KIP2, USH1J |
Location: |
Chr.15:78104606-78131976 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB2 | NM_001271888.1 | 4 | 710 | NP_001258817.1 |
| NM_001271889.1 | 3 | 598 | NP_001258818.1 | |
| NM_001301224.1 | 4 | 760 | NP_001288153.1 | |
| NM_006383.3 | 5 | 745 | NP_006374.1 | |
| NR_125435.1 | 4 | 633 | ||
| XM_005254126.3 | 5 | 1186 | XP_005254183.1 | |
| XM_006720374.2 | 6 | 570 | XP_006720437.1 | |
| XM_011521161.2 | 5 | 454 | XP_011519463.1 | |
| AB012955.1 | 5 | 431 | BAA33584.1 | |
| AI817087.1 | 4 | 369 | ||
| AI857912.1 | ||||
| AK293167.1 | 5 | 659 | ||
| AL110098.1 | ||||
| BC033108.1 | 4 | 710 | ||
| BC047381.1 | 5 | 673 | AAH47381.1 | |
| BE042882.1 | 5 | 481 | ||
| BI523557.1 | 2 | 145 | ||
| BT019962.1 | 5 | 425 | AAV38765.1 | |
| BU556614.1 | 3 | 496 | ||
| CN371149.1 | ||||
| CN371150.1 | 3 | 382 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv274890 | Chr.15:78104105 - 78109916 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map