Assay Details
Target Gene Details
| Entrez Gene ID: | 4709 | 
| Gene Name: | NADH:ubiquinone oxidoreductase subunit B3 | 
| Gene Aliases: | B12, CI-B12 | 
| Location: | Chr.2:201071739-201085750 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NDUFB3 | NM_001257102.1 | 1 | 263 | NP_001244031.1 | 
| NM_002491.2 | 1 | 263 | NP_002482.1 | |
| XM_011511230.2 | 1 | 217 | XP_011509532.1 | |
| AF035839.1 | AAC15590.1 | |||
| AF047183.1 | 1 | 191 | AAC04268.1 | |
| BC018183.1 | 1 | 211 | AAH18183.1 | |
| BM848639.1 | 1 | 263 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv584160 | Chr.2:201039216 - 201298760 on Build GRCh38 | Loss |  CFLAR-AS1  NDUFB3  CASP8  FAM126B  RPL17P10  ALS2CR12  CFLAR  CASP10 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top