Assay Details
Target Gene Details
Entrez Gene ID: | 148932 |
Gene Name: | MOB kinase activator 3C |
Gene Aliases: |
MOB1E, MOBKL2C |
Location: |
Chr.1:46607715-46616891 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB3C | NM_145279.4 | 4 | 2387 | NP_660322.2 |
| NM_201403.2 | 4 | 2405 | NP_958805.1 | |
| XM_017000407.1 | 3 | 5336 | XP_016855896.1 | |
| AK090453.1 | 3 | 5318 | ||
| AK091808.1 | 3 | 2131 | BAC03752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009193 | Chr.1:46474327 - 46619995 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map