Assay Details
Target Gene Details
Entrez Gene ID: | 51360 |
Gene Name: | membrane bound transcription factor peptidase, site 2 |
Gene Aliases: |
BRESEK, IFAP, KFSD, KFSDX, OLMSX, S2P |
Location: |
Chr.X:21839538-21885423 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBTPS2 | NM_015884.3 | 6 | 921 | NP_056968.1 |
| AF019612.1 | 6 | 823 | AAC51937.1 | |
| AK292933.1 | 6 | 828 | ||
| AK307143.1 | 2 | 192 | ||
| AK312474.1 | 6 | 754 |
More Information
Set Membership: |
|

Genomic Map