Assay Details
Target Gene Details
Entrez Gene ID: | 3171 |
Gene Name: | forkhead box A3 |
Gene Aliases: |
FKHH3, HNF3G, TCF3G |
Location: |
Chr.19:45864260-45873797 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXA3 | NM_004497.2 | 2 | 1234 | NP_004488.2 |
| AK223473.1 | 2 | 1163 | BAD97193.1 | |
| BC016024.1 | 2 | 1234 | AAH16024.1 | |
| BT006720.1 | AAP35366.1 | |||
| L12141.1 | AAA58477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510770 | Chr.19:45841909 - 45921434 on Build GRCh38 | Deletion |
|
| nsv523029 | Chr.19:45872959 - 45876389 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map