Assay Details
Target Gene Details
| Entrez Gene ID: | 4791 | 
| Gene Name: | nuclear factor kappa B subunit 2 | 
| Gene Aliases: | CVID10, H2TF1, LYT-10, LYT10, NF-kB2, p100, p49/p100, p52 | 
| Location: | Chr.10:102394110-102402529 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Exon 4 - Intron 4 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NFKB2 | NM_001077494.3 | NP_001070962.1 | ||
| NM_001261403.2 | NP_001248332.1 | |||
| NM_001288724.1 | NP_001275653.1 | |||
| NM_001322934.1 | NP_001309863.1 | |||
| NM_001322935.1 | NP_001309864.1 | |||
| NM_002502.5 | NP_002493.3 | |||
| XM_011539830.2 | XP_011538132.1 | |||
| XM_017016278.1 | XP_016871767.1 | |||
| AK292654.1 | ||||
| BC002844.2 | AAH02844.1 | |||
| BT009769.1 | AAP88771.1 | |||
| BY799958.2 | ||||
| DA349980.1 | ||||
| DA408461.1 | ||||
| DA711063.1 | ||||
| S76638.1 | AAB21124.1 | |||
| U09609.2 | AAA21462.2 | |||
| X61498.1 | CAA43715.1 | |||
| X61499.1 | CAA43716.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1159761 | Chr.10:102372778 - 102430809 on Build GRCh38 | Deletion |  CUEDC2  FBXL15  PSD  GBF1  NFKB2 | 
| nsv526665 | Chr.10:102363085 - 102397954 on Build GRCh38 | Loss |  GBF1  NFKB2 | 
| nsv525546 | Chr.10:102380593 - 102415138 on Build GRCh38 | Loss |  PSD  GBF1  NFKB2 | 
| nsv951006 | Chr.10:102394644 - 102425743 on Build GRCh38 | Deletion |  CUEDC2  FBXL15  PSD  NFKB2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top