Assay Details
Target Gene Details
| Entrez Gene ID: | 7075 | 
| Gene Name: | tyrosine kinase with immunoglobulin like and EGF like domains 1 | 
| Gene Aliases: | JTK14, TIE | 
| Location: | Chr.1:43300895-43323110 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Exon 3 - Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| TIE1 | NM_001253357.1 | NP_001240286.1 | ||
| NM_005424.4 | NP_005415.1 | |||
| XM_005271163.1 | XP_005271220.1 | |||
| XM_006710869.1 | XP_006710932.1 | |||
| XM_017002207.1 | XP_016857696.1 | |||
| AF217976.1 | AAG17219.1 | |||
| AK225560.1 | ||||
| AK300399.1 | ||||
| AK300623.1 | ||||
| AK307639.1 | ||||
| BC038239.1 | AAH38239.1 | |||
| EU826586.1 | ||||
| EU826587.1 | ||||
| EU826588.1 | ||||
| EU826590.1 | ||||
| X60957.1 | CAA43290.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |  TIE1  CDC20  HYI  SZT2  MPL  MIR6735  MED8  ELOVL1  MIR6734 | 
| nsv829693 | Chr.1:43129457 - 43312977 on Build GRCh38 | Loss |  CFAP57  TMEM125  TIE1  C1orf210  MIR6733  FAM183A  EBNA1BP2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top