Assay Details
Target Gene Details
Entrez Gene ID: | 9746 |
Gene Name: | calsyntenin 3 |
Gene Aliases: |
CDHR14, CSTN3, alcbeta |
Location: |
Chr.12:7130371-7158945 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLSTN3 | NM_014718.3 | 3 | 577 | NP_055533.2 |
| XM_006719163.3 | 2 | 1903 | XP_006719226.1 | |
| AB018269.1 | 2 | 873 | BAA34446.2 | |
| AJ277460.1 | 3 | 574 | CAC33084.1 | |
| AK299177.1 | 3 | 551 | ||
| AK316059.1 | 3 | 577 | ||
| AY753302.1 | 2 | 335 | AAV30552.1 | |
| BC104767.1 | 3 | 381 | AAI04768.1 | |
| BC111491.1 | 3 | 565 | AAI11492.1 | |
| BC112283.1 | 3 | 393 | AAI12284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818882 | Chr.12:7128721 - 7149386 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map