Assay Details
Target Gene Details
Entrez Gene ID: | 131096 |
Gene Name: | potassium voltage-gated channel subfamily H member 8 |
Gene Aliases: |
ELK, ELK1, Kv12.1, elk3 |
Location: |
Chr.3:19148454-19535643 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH8 | NM_144633.2 | 1 | 159 | NP_653234.2 |
| XM_017005698.1 | 1 | 230 | XP_016861187.1 | |
| XM_017005699.1 | 1 | 130 | XP_016861188.1 | |
| XM_017005700.1 | 1 | 230 | XP_016861189.1 | |
| XM_017005701.1 | 1 | 217 | XP_016861190.1 | |
| XM_017005702.1 | 1 | 130 | XP_016861191.1 | |
| AB209053.1 | 1 | 150 | BAD92290.1 | |
| AK295633.1 | 1 | 206 | ||
| AK309034.1 | 1 | 135 | ||
| AY053503.1 | 1 | 159 | AAL15429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010123 | Chr.3:18948815 - 19948012 on Build GRCh38 | Gain |
|
| nsv517847 | Chr.3:18803855 - 19442203 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map