Assay Details
Target Gene Details
Entrez Gene ID: | 2488 |
Gene Name: | follicle stimulating hormone beta subunit |
Gene Aliases: |
HH24 |
Location: |
Chr.11:30231016-30235277 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSHB | NM_000510.2 | 3 | 1220 | NP_000501.1 |
| NM_001018080.1 | 3 | 1190 | NP_001018090.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553946 | Chr.11:30194611 - 30260631 on Build GRCh38 | Loss |
|
| nsv437712 | Chr.11:30143725 - 30332178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map