Assay Details
Target Gene Details
Entrez Gene ID: | 4085 |
Gene Name: | MAD2 mitotic arrest deficient-like 1 (yeast) |
Gene Aliases: |
HSMAD2, MAD2 |
Location: |
Chr.4:120059422-120066858 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAD2L1 | NM_002358.3 | NP_002349.1 | ||
| AA443733.1 | ||||
| AF394735.1 | AAN74648.1 | |||
| AJ000186.1 | CAA03943.1 | |||
| AK223433.1 | BAD97153.1 | |||
| AK298228.1 | ||||
| AK313827.1 | ||||
| BC000356.2 | AAH00356.1 | |||
| BC005945.1 | AAH05945.1 | |||
| BC070283.1 | AAH70283.1 |
More Information
Set Membership: |
|

Genomic Map