Assay Details
Target Gene Details
Entrez Gene ID: | 2894 |
Gene Name: | glutamate ionotropic receptor delta type subunit 1 |
Gene Aliases: |
GluD1 |
Location: |
Chr.10:85599555-86366493 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRID1 | NM_017551.2 | 16 | 4685 | NP_060021.1 |
| AB033046.1 | 13 | 3932 | BAA86534.1 |
Target Gene Details
Entrez Gene ID: | 100507470 |
Gene Name: | GRID1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:85577731-85607215 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRID1-AS1 | NR_038986.1 | |||
| AK097655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891878 | Chr.10:85560815 - 85628886 on Build GRCh38 | Loss |
|
| nsv1052427 | Chr.10:85234813 - 86270793 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map