Assay Details
Target Gene Details
Entrez Gene ID: | 10006 |
Gene Name: | abl interactor 1 |
Gene Aliases: |
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1 |
Location: |
Chr.10:26746596-26861087 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABI1 | NM_001012750.2 | 11 | 2655 | NP_001012768.1 |
| NM_001012751.2 | 10 | 2568 | NP_001012769.1 | |
| NM_001012752.2 | 10 | 2640 | NP_001012770.1 | |
| NM_001178116.1 | 12 | 2700 | NP_001171587.1 | |
| NM_001178119.1 | 11 | 2652 | NP_001171590.1 | |
| NM_001178120.1 | 10 | 2565 | NP_001171591.1 | |
| NM_001178121.1 | 9 | 2550 | NP_001171592.1 | |
| NM_001178122.1 | 10 | 2478 | NP_001171593.1 | |
| NM_001178123.1 | 9 | 2391 | NP_001171594.1 | |
| NM_001178124.1 | 8 | 2376 | NP_001171595.1 | |
| NM_001178125.1 | 7 | 2199 | NP_001171596.1 | |
| NM_005470.3 | 12 | 2736 | NP_005461.2 | |
| XM_005252332.3 | 11 | 2536 | XP_005252389.1 | |
| XM_005252333.3 | 10 | 2452 | XP_005252390.1 | |
| XM_006717359.2 | 12 | 2632 | XP_006717422.1 | |
| XM_006717360.2 | 11 | 2548 | XP_006717423.1 | |
| XM_006717361.2 | 9 | 2368 | XP_006717424.1 | |
| XM_017015458.1 | 11 | 2545 | XP_016870947.1 | |
| AB209268.1 | 10 | 2416 | BAD92505.1 | |
| AF006516.1 | 11 | 2494 | AAB62569.1 | |
| AF260262.1 | AAF70309.1 | |||
| AK126803.1 | 12 | 2599 | ||
| BC024254.2 | 10 | 2491 | AAH24254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3415799 | Chr.10:26640952 - 26944612 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map