Assay Details
Target Gene Details
Entrez Gene ID: | 57139 |
Gene Name: | ral guanine nucleotide dissociation stimulator like 3 |
Gene Aliases: |
- |
Location: |
Chr.19:11394056-11419342 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGL3 | NM_001035223.3 | 7 | 884 | NP_001030300.3 |
| NM_001161616.2 | 7 | 884 | NP_001155088.2 | |
| AK057147.1 | 8 | 1039 | ||
| AK096811.1 | 3 | 201 | ||
| AK293404.1 | 6 | 611 | ||
| BC014426.1 | 7 | 872 | ||
| BC101756.1 | 7 | 870 | AAI01757.1 | |
| BC111958.1 | 7 | 870 | AAI11959.1 | |
| BC143530.1 | 7 | 870 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833752 | Chr.19:11208324 - 11448939 on Build GRCh38 | Loss |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map