Assay Details
Target Gene Details
Entrez Gene ID: | 3782 |
Gene Name: | potassium calcium-activated channel subfamily N member 3 |
Gene Aliases: |
KCa2.3, SK3, SKCA3, hSK3 |
Location: |
Chr.1:154697462-154870278 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNN3 | NM_001204087.1 | NP_001191016.1 | ||
| NM_002249.5 | NP_002240.3 | |||
| NM_170782.2 | NP_740752.1 | |||
| AF031815.1 | AAC26099.1 | |||
| AF438203.1 | AAL40801.1 | |||
| AJ251016.1 | CAB61331.1 | |||
| AY049734.1 | AAL10706.1 | |||
| AY138900.1 | AAN46636.1 | |||
| AY258142.1 | AAP45947.1 | |||
| BC042147.1 | AAH42147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509491 | Chr.1:154692639 - 154719309 on Build GRCh38 | Insertion |
|
| nsv547958 | Chr.1:154596207 - 154828771 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map