Assay Details
Target Gene Details
Entrez Gene ID: | 57110 |
Gene Name: | HRAS like suppressor |
Gene Aliases: |
A-C1, H-REV107, HRASLS1, HRSL1, HSD28 |
Location: |
Chr.3:193241128-193281639 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRASLS | NM_020386.4 | 3 | 594 | NP_065119.2 |
| XM_011513034.1 | 3 | 594 | XP_011511336.1 | |
| XM_011513035.2 | 3 | 594 | XP_011511337.1 | |
| XM_017006923.1 | 4 | 475 | XP_016862412.1 | |
| AB030816.1 | 3 | 399 | BAB08110.1 | |
| AB510981.1 | 2 | 188 | ||
| AY251533.1 | 2 | 201 | AAP20056.1 | |
| BC048095.1 | 3 | 597 | AAH48095.1 | |
| BX098792.1 | 2 | 483 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003505 | Chr.3:193168634 - 193574329 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map