Assay Details
Target Gene Details
Entrez Gene ID: | 79174 |
Gene Name: | cysteine rich with EGF like domains 2 |
Gene Aliases: |
- |
Location: |
Chr.22:49918630-49927540 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRELD2 | NM_001135101.2 | 10 | 1227 | NP_001128573.1 |
| NM_001284317.1 | 8 | 996 | NP_001271246.1 | |
| NM_001284318.1 | 8 | 984 | NP_001271247.1 | |
| NM_024324.4 | 9 | 1080 | NP_077300.3 | |
| NR_104295.1 | 9 | 1223 | ||
| XM_005261737.2 | 10 | 1218 | XP_005261794.1 | |
| XM_005261738.4 | 9 | 1074 | XP_005261795.1 | |
| XM_017028938.1 | 9 | 1004 | XP_016884427.1 | |
| AK126178.1 | 1 | 2104 | ||
| AK225930.1 | 9 | 1005 | ||
| AK308692.1 | 9 | 1216 | ||
| AL713788.1 | 1 | 2279 | CAH56485.1 | |
| AY358355.1 | 9 | 1004 | AAQ88721.1 | |
| BC002894.1 | 8 | 887 | AAH02894.1 | |
| BC050675.1 | 9 | 1014 | AAH50675.1 | |
| BQ003113.1 | 2 | 371 | ||
| DQ470676.1 | 8 | 887 | ||
| DQ470677.1 | 8 | 899 | ||
| DQ470678.1 | 9 | 1014 | ||
| DQ470679.1 | 10 | 1222 | ||
| DQ470680.1 | 9 | 983 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1334e212 | Chr.22:49920870 - 49933235 on Build GRCh38 | Loss |
|
| nsv471221 | Chr.22:49880243 - 49927295 on Build GRCh38 | Loss |
|
| dgv1335e212 | Chr.22:49924410 - 49933235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map