Assay Details
Target Gene Details
Entrez Gene ID: | 55715 |
Gene Name: | docking protein 4 |
Gene Aliases: |
IRS-5, IRS5 |
Location: |
Chr.16:57471958-57486473 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK4 | NM_018110.3 | 2 | 358 | NP_060580.2 |
| XM_005256042.3 | 1 | 300 | XP_005256099.1 | |
| XM_005256043.2 | 1 | 300 | XP_005256100.1 | |
| AF466369.1 | AAL74195.1 | |||
| AK001350.1 | 2 | 285 | BAA91642.1 | |
| AK307270.1 | 1 | 257 | ||
| AK315426.1 | 1 | 268 | ||
| AW163719.1 | ||||
| BC001540.2 | 1 | 57 | AAH01540.1 | |
| BC003541.1 | 2 | 300 | AAH03541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map