Assay Details
Target Gene Details
Entrez Gene ID: | 338645 |
Gene Name: | leucine zipper protein 2 |
Gene Aliases: |
KFSP2566, PRO6246 |
Location: |
Chr.11:24494304-25082640 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP2 | NM_001009909.3 | 1 | 234 | NP_001009909.2 |
| NM_001252008.1 | 1 | 234 | NP_001238937.1 | |
| NM_001252010.1 | 1 | 234 | NP_001238939.1 | |
| XM_011520056.2 | 1 | 2894 | XP_011518358.1 | |
| XM_017017648.1 | 1 | 151 | XP_016873137.1 | |
| XM_017017649.1 | 1 | 151 | XP_016873138.1 | |
| XM_017017650.1 | 1 | 234 | XP_016873139.1 | |
| XM_017017651.1 | 1 | 234 | XP_016873140.1 | |
| AK127695.1 | 1 | 234 | BAC87089.1 | |
| AK309077.1 | ||||
| AL832641.3 | 1 | 146 | CAD90000.1 | |
| AY164460.1 | 1 | 194 | ||
| AY358386.1 | 1 | 65 | AAQ88752.1 | |
| BC132829.1 | ||||
| DA706662.1 | 1 | 234 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553819 | Chr.11:24477824 - 24523807 on Build GRCh38 | Loss |
|
| nsv1040177 | Chr.11:23010756 - 24853532 on Build GRCh38 | Gain |
|
| nsv467755 | Chr.11:24385835 - 24506412 on Build GRCh38 | Loss |
|
| nsv469945 | Chr.11:24493977 - 24523807 on Build GRCh38 | Gain |
|

Genomic Map