Assay Details
Target Gene Details
Entrez Gene ID: | 28956 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 2 |
Gene Aliases: |
ENDAP, HSPC003, MAPBPIP, MAPKSP1AP, ROBLD3, Ragulator2, p14 |
Location: |
Chr.1:156054726-156058510 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR2 | NM_001145264.1 | NP_001138736.1 | ||
| NM_014017.3 | NP_054736.1 | |||
| AF070659.1 | AAD20965.1 | |||
| AK307086.1 | ||||
| BC024190.2 | AAH24190.1 | |||
| BM353842.1 | ||||
| BU184679.1 | ||||
| CA438137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509513 | Chr.1:155932322 - 156178790 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map