Assay Details
Target Gene Details
Entrez Gene ID: | 10936 |
Gene Name: | G protein-coupled receptor 75 |
Gene Aliases: |
GPRchr2, WI31133 |
Location: |
Chr.2:53852913-53860033 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR75 | NM_006794.3 | 2 | 325 | NP_006785.1 |
| AF072693.1 | 2 | 220 | AAD19849.1 | |
| AK314885.1 | 2 | 281 | ||
| BC067475.1 | 1 | 121 | AAH67475.1 | |
| BC067476.1 | AAH67476.1 |
Target Gene Details
Entrez Gene ID: | 100302652 |
Gene Name: | GPR75-ASB3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:53670293-53860033 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR75-ASB3 | NM_001164165.1 | NP_001157637.1 | ||
| DC314536.1 |
More Information
Set Membership: |
|

Genomic Map