Assay Details
Target Gene Details
Entrez Gene ID: | 4838 |
Gene Name: | nodal growth differentiation factor |
Gene Aliases: |
HTX5 |
Location: |
Chr.10:70431936-70441709 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NODAL | NM_018055.4 | 3 | 1530 | NP_060525.3 |
| BC033585.1 | 3 | 1424 | AAH33585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77232076] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map