Assay Details
Target Gene Details
Entrez Gene ID: | 1992 |
Gene Name: | serpin family B member 1 |
Gene Aliases: |
EI, ELANH2, HEL-S-27, HEL57, LEI, M/NEI, MNEI, PI-2, PI2 |
Location: |
Chr.6:2832332-2842049 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB1 | NM_030666.3 | NP_109591.1 | ||
| NR_073111.1 | ||||
| NR_073112.1 | ||||
| XM_011514333.1 | XP_011512635.1 | |||
| XM_011514334.2 | XP_011512636.1 | |||
| XM_011514335.2 | XP_011512637.1 | |||
| AK223370.1 | BAD97090.1 | |||
| AK291327.1 | ||||
| AK298044.1 | ||||
| AK304641.1 | ||||
| AK309765.1 | ||||
| AK312973.1 | ||||
| BC009015.1 | AAH09015.1 | |||
| BT006928.1 | AAP35574.1 | |||
| BU942473.1 | ||||
| EU794624.1 | ||||
| GQ472232.1 | ||||
| M93056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600831 | Chr.6:2803955 - 2843101 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map