Assay Details
Target Gene Details
Entrez Gene ID: | 84286 |
Gene Name: | transmembrane protein 175 |
Gene Aliases: |
hTMEM175 |
Location: |
Chr.4:932387-958656 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM175 | NM_001297423.1 | 11 | 1629 | NP_001284352.1 |
| NM_001297424.1 | 9 | 945 | NP_001284353.1 | |
| NM_001297425.1 | 9 | 942 | NP_001284354.1 | |
| NM_001297426.1 | 12 | 1409 | NP_001284355.1 | |
| NM_001297427.1 | 11 | 1665 | NP_001284356.1 | |
| NM_001297428.1 | 9 | 981 | NP_001284357.1 | |
| NM_032326.3 | 11 | 1168 | NP_115702.1 | |
| XM_005272301.2 | 10 | 1059 | XP_005272358.1 | |
| XM_005272303.4 | 12 | 1325 | XP_005272360.1 | |
| XM_005272304.1 | 11 | 1134 | XP_005272361.1 | |
| XM_011513576.1 | 7 | 686 | XP_011511878.1 | |
| XM_017008701.1 | 12 | 1245 | XP_016864190.1 | |
| XM_017008702.1 | 9 | 892 | XP_016864191.1 | |
| XM_017008703.1 | 12 | 1687 | XP_016864192.1 | |
| XM_017008704.1 | 10 | 1500 | XP_016864193.1 | |
| XM_017008705.1 | 8 | 837 | XP_016864194.1 | |
| AL834199.1 | 12 | 1279 | CAD38888.1 | |
| BC005158.2 | 11 | 1081 | AAH05158.1 | |
| CF242841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003219 | Chr.4:776106 - 1111676 on Build GRCh38 | Gain |
|
| nsv593226 | Chr.4:948218 - 1025223 on Build GRCh38 | Loss |
|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| nsv950327 | Chr.4:945913 - 978512 on Build GRCh38 | Deletion |
|
| dgv5079n100 | Chr.4:843485 - 970603 on Build GRCh38 | Gain |
|
| esv29984 | Chr.4:857569 - 970571 on Build GRCh38 | Loss |
|
| nsv469998 | Chr.4:815805 - 1093743 on Build GRCh38 | Loss |
|
| nsv1008352 | Chr.4:819605 - 1041628 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map