Assay Details
Target Gene Details
Entrez Gene ID: | 582 |
Gene Name: | Bardet-Biedl syndrome 1 |
Gene Aliases: |
BBS2L2 |
Location: |
Chr.11:66510648-66533613 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBS1 | NM_024649.4 | 17 | 2390 | NP_078925.3 |
| AF503941.1 | 17 | 2390 | AAM92770.1 | |
| AK027243.1 | 5 | 1087 | BAB15704.1 | |
| AK095638.1 | 15 | 2605 | ||
| BC047642.1 | 16 | 2467 | AAH47642.1 | |
| BX647612.1 | 17 | 2419 | CAI46060.1 |
Target Gene Details
Entrez Gene ID: | 254359 |
Gene Name: | zinc finger DHHC-type containing 24 |
Gene Aliases: |
- |
Location: |
Chr.11:66520625-66546200 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC24 | XM_005273874.3 | XP_005273931.1 | ||
| XM_011544894.1 | XP_011543196.1 | |||
| AK310561.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map