Assay Details
Target Gene Details
Entrez Gene ID: | 5887 |
Gene Name: | RAD23 homolog B, nucleotide excision repair protein |
Gene Aliases: |
HHR23B, HR23B, P58 |
Location: |
Chr.9:107283236-107332194 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD23B | NM_001244713.1 | NP_001231642.1 | ||
| NM_001244724.1 | NP_001231653.1 | |||
| NM_002874.4 | NP_002865.1 | |||
| AK125226.1 | ||||
| AK223479.1 | BAD97199.1 | |||
| AK293532.1 | ||||
| AK297986.1 | ||||
| AK316189.1 | ||||
| AL540969.3 | ||||
| AY313777.1 | AAP81008.1 | |||
| BC020973.2 | AAH20973.1 | |||
| BI460482.1 | ||||
| D21090.1 | BAA04652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831682 | Chr.9:107174900 - 107313172 on Build GRCh38 | Gain |
|
| nsv519266 | Chr.9:107287196 - 107322974 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map