Assay Details
Target Gene Details
Entrez Gene ID: | 4610 |
Gene Name: | v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog |
Gene Aliases: |
L-Myc, LMYC, MYCL1, bHLHe38 |
Location: |
Chr.1:39895424-39902015 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCL | NM_001033081.2 | 1 | 363 | NP_001028253.1 |
| NM_001033082.2 | NP_001028254.2 | |||
| NM_005376.4 | NP_005367.2 | |||
| AK296078.1 | ||||
| BC011864.2 | AAH11864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map