| nsv470247 | 
						
							
								Chr.8:143283403 - 143615483
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 EEF1D
							
								 GSDMD
							
								 RHPN1-AS1
							
								 NAPRT
							
								 MAFA-AS1
							
								 MROH6
							
								 TIGD5
							
								 ZC3H3
							
								 ZNF696
							
								 PYCRL
							
								 TSTA3
							
								 TOP1MT
							
								 MAFA
							
								 RHPN1
							
						 | 
					
				
		    		
						| nsv1161810 | 
						
							
								Chr.8:143542268 - 143622987
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZC3H3
							
								 GSDMD
							
								 PYCRL
							
								 TSTA3
							
								 NAPRT
							
								 MROH6
							
						 | 
					
				
		    		
						| nsv612793 | 
						
							
								Chr.8:143552167 - 143630578
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 EEF1D
							
								 TIGD5
							
								 GSDMD
							
								 PYCRL
							
								 TSTA3
							
								 NAPRT
							
								 MROH6
							
						 | 
					
				
		    		
						| nsv516210 | 
						
							
								Chr.8:143533025 - 143615483
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZC3H3
							
								 GSDMD
							
								 PYCRL
							
								 TSTA3
							
								 NAPRT
							
								 MROH6
							
						 | 
					
				
		    		
						| nsv950013 | 
						
							
								Chr.8:143465031 - 143616230
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZC3H3
							
								 GSDMD
							
								 PYCRL
							
								 TSTA3
							
								 NAPRT
							
								 MROH6
							
						 | 
					
				
		    		
						| nsv612777 | 
						
							
								Chr.8:143538914 - 143632493
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZC3H3
							
								 GSDMD
							
								 PYCRL
							
								 TSTA3
							
								 NAPRT
							
								 MROH6
							
						 | 
					
				
		    		
						| esv3584899 | 
						
							
								Chr.8:143611680 - 143636819
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 ZNF623
							
								 TSTA3
							
						 | 
					
				
		    		
						| nsv42 | 
						
							
								Chr.8:143610600 - 143635500
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 TSTA3
							
						 | 
					
				
		    		
						| nsv6439 | 
						
							
								Chr.8:143606519 - 143651282
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 ZNF623
							
								 PYCRL
							
								 TSTA3
							
						 | 
					
				
		    		
						| dgv7336n100 | 
						
							
								Chr.8:143510004 - 143616975
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZC3H3
							
								 GSDMD
							
								 PYCRL
							
								 TSTA3
							
								 NAPRT
							
								 MROH6
							
						 | 
					
				
		    		
						| dgv271n111 | 
						
							
								Chr.8:143601764 - 143642893
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 ZNF623
							
								 PYCRL
							
								 TSTA3
							
						 | 
					
				
		    		
						| nsv831488 | 
						
							
								Chr.8:143590454 - 143684444
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZNF623
							
								 PYCRL
							
								 TSTA3
							
						 | 
					
				
		    		
						| nsv466004 | 
						
							
								Chr.8:143581491 - 143640022
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 EEF1D
							
								 TIGD5
							
								 ZNF623
							
								 PYCRL
							
								 TSTA3
							
						 |