Assay Details
Target Gene Details
Entrez Gene ID: | 10459 |
Gene Name: | MAD2 mitotic arrest deficient-like 2 (yeast) |
Gene Aliases: |
MAD2B, POLZ2, REV7 |
Location: |
Chr.1:11674480-11691788 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAD2L2 | NM_001127325.1 | NP_001120797.1 | ||
| NM_006341.3 | NP_006332.3 | |||
| XM_011540507.1 | XP_011538809.1 | |||
| AF072933.1 | AAD41647.1 | |||
| AF080398.1 | AAF20267.1 | |||
| AF139365.1 | AAD30290.1 | |||
| AF157482.1 | AAF34357.1 | |||
| AK027327.1 | ||||
| AK094316.1 | ||||
| BC015244.1 | AAH15244.1 | |||
| BI517654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545416 | Chr.1:11620226 - 11676353 on Build GRCh38 | Loss |
|
| nsv832758 | Chr.1:11637026 - 11777087 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map