Assay Details
Target Gene Details
Entrez Gene ID: | 54474 |
Gene Name: | keratin 20 |
Gene Aliases: |
CD20, CK-20, CK20, K20, KRT21 |
Location: |
Chr.17:40875889-40885243 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT20 | NM_019010.2 | NP_061883.1 | ||
| AK312744.1 | ||||
| BC031559.1 | AAH31559.1 | |||
| X73502.1 | CAA51914.1 |
More Information
Set Membership: |
|

Genomic Map