Assay Details
Target Gene Details
Entrez Gene ID: | 57699 |
Gene Name: | copine 5 |
Gene Aliases: |
COPN5, CPN5 |
Location: |
Chr.6:36740773-36839444 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPNE5 | NM_001314018.1 | 10 | 1688 | NP_001300947.1 |
| NM_001314019.1 | 11 | 1729 | NP_001300948.1 | |
| NM_001314020.1 | 9 | 1626 | NP_001300949.1 | |
| NM_020939.1 | 21 | 2485 | NP_065990.1 | |
| XM_005249247.2 | 22 | 2536 | XP_005249304.1 | |
| AB046819.1 | 21 | 3043 | ||
| AL833002.1 | 1 | 1965 | CAH56298.1 | |
| BC053872.1 | 10 | 1688 | AAH53872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524053 | Chr.6:36727446 - 36779629 on Build GRCh38 | Loss |
|
| nsv602951 | Chr.6:36738380 - 36751302 on Build GRCh38 | Loss |
|
| dgv379n21 | Chr.6:36734507 - 36745943 on Build GRCh38 | Loss |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map