Assay Details
Target Gene Details
Entrez Gene ID: | 4684 |
Gene Name: | neural cell adhesion molecule 1 |
Gene Aliases: |
CD56, MSK39, NCAM |
Location: |
Chr.11:112961247-113278436 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAM1 | NM_000615.6 | 17 | 2582 | NP_000606.3 |
| NM_001242607.1 | 19 | 2690 | NP_001229536.1 | |
| NM_181351.4 | 18 | 2612 | NP_851996.2 | |
| AK057509.1 | 17 | 2685 | ||
| AK292453.1 | 19 | 2247 | ||
| AK299865.1 | 8 | 793 | ||
| AK314589.1 | 19 | 2662 | ||
| BC014205.2 | 12 | 1476 | AAH14205.2 | |
| BC047244.1 | 18 | 2424 | AAH47244.1 | |
| M17410.1 | 6 | 743 | AAA59913.1 | |
| S71824.1 | 17 | 2417 | AAB31836.1 | |
| U63041.1 | 17 | 2253 | AAB04558.1 | |
| X55322.1 | 2 | 163 |
Target Gene Details
Entrez Gene ID: | 100288346 |
Gene Name: | NCAM1 antisense RNA1 |
Gene Aliases: |
- |
Location: |
Chr.11:113269532-113273901 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAM1-AS1 | NR_034101.1 | 2 | 498 | |
| AK293718.1 | 2 | 429 | ||
| DC305174.1 | 2 | 349 | ||
| DC309426.1 | 2 | 498 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483080 | Chr.11:113216072 - 113400474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map