Assay Details
Target Gene Details
Entrez Gene ID: | 55370 |
Gene Name: | protein phosphatase 4 regulatory subunit 1 like (pseudogene) |
Gene Aliases: |
C20orf192, PPP4R1P1, PRO1085 |
Location: |
Chr.20:58232777-58309439 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R1L | NR_003505.3 | 10 | 1406 | |
| AK057583.1 | 10 | 1406 | BAB71529.1 | |
| AL833409.1 | 7 | 2369 | ||
| CX783537.1 | 2 | 392 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055956 | Chr.20:58211957 - 58367256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map