Assay Details
Target Gene Details
Entrez Gene ID: | 6144 |
Gene Name: | ribosomal protein L21 |
Gene Aliases: |
HYPT12, L21 |
Location: |
Chr.13:27251555-27256568 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL21 |
Target Gene Details
Entrez Gene ID: | 619499 |
Gene Name: | small nucleolar RNA, H/ACA box 27 |
Gene Aliases: |
ACA27 |
Location: |
Chr.13:27255401-27255526 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNORA27 | NR_002575.1 | |||
| AJ609439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976098 | Chr.13:27253748 - 27256597 on Build GRCh38 | Duplication |
|

Genomic Map