Assay Details
Target Gene Details
Entrez Gene ID: | 64800 |
Gene Name: | EF-hand calcium binding domain 6 |
Gene Aliases: |
DJBP, HSCBCIP1, dJ185D5.1 |
Location: |
Chr.22:43528549-43812403 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 37 - Exon 37 |
Target Gene Details
Entrez Gene ID: | 100874197 |
Gene Name: | EFCAB6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:43516254-43536605 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB6-AS1 | NR_046563.1 | |||
| AK090843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058659 | Chr.22:43522577 - 43700374 on Build GRCh38 | Loss |
|
| nsv589098 | Chr.22:43463583 - 43718462 on Build GRCh38 | Gain |
|
| nsv471205 | Chr.22:43467417 - 43718981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map