Assay Details
Target Gene Details
Entrez Gene ID: | 283461 |
Gene Name: | chromosome 12 open reading frame 40 |
Gene Aliases: |
HEL-206, HEL-S-94 |
Location: |
Chr.12:39626167-39908300 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf40 | NR_135051.1 | |||
| AK097445.1 | BAC05057.1 | |||
| EU668334.1 | ||||
| GQ472216.1 |
Target Gene Details
Entrez Gene ID: | 114134 |
Gene Name: | solute carrier family 2 member 13 |
Gene Aliases: |
HMIT |
Location: |
Chr.12:39755021-40106085 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A13 | NM_052885.3 | 10 | 1986 | NP_443117.3 |
| XM_011537847.2 | 9 | 2086 | XP_011536149.1 | |
| XM_017018764.1 | 10 | 1459 | XP_016874253.1 | |
| XM_017018765.1 | 10 | 2097 | XP_016874254.1 | |
| XM_017018766.1 | 9 | 1244 | XP_016874255.1 | |
| AJ315644.1 | 10 | 1986 | CAC51116.1 | |
| BC117117.1 | 10 | 1930 | ||
| BC117119.1 | 10 | 1930 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428280 | Chr.12:39728911 - 39805819 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map