Assay Details
Target Gene Details
Entrez Gene ID: | 5655 |
Gene Name: | kallikrein related peptidase 10 |
Gene Aliases: |
NES1, PRSSL1 |
Location: |
Chr.19:51012744-51020175 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK10 | NM_001077500.1 | 6 | 883 | NP_001070968.1 |
| NM_002776.4 | 6 | 982 | NP_002767.2 | |
| NM_145888.2 | 6 | 844 | NP_665895.1 | |
| XM_005259061.3 | 6 | 904 | XP_005259118.1 | |
| XM_005259062.3 | 6 | 868 | XP_005259119.1 | |
| XM_006723287.3 | 6 | 871 | XP_006723350.1 | |
| XM_006723289.3 | 6 | 966 | XP_006723352.1 | |
| XM_017026993.1 | 6 | 881 | XP_016882482.1 | |
| AF024605.1 | 6 | 844 | AAB81602.1 | |
| AK292365.1 | 6 | 973 | ||
| AK309456.1 | 5 | 1070 | ||
| AY561635.1 | 6 | 911 | AAS66976.1 | |
| BC002710.2 | 6 | 982 | AAH02710.1 | |
| CR450327.1 | 5 | 763 | CAG29323.1 | |
| KJ010190.1 | ||||
| KJ010191.1 | ||||
| KJ010192.1 | ||||
| KJ010193.1 | ||||
| KJ010194.1 | ||||
| KJ010195.1 | ||||
| KJ010196.1 | ||||
| KJ010197.1 | ||||
| KJ010198.1 | ||||
| KJ010199.1 | ||||
| KJ010200.1 | ||||
| KJ010201.1 | ||||
| KJ010202.1 |
More Information
Set Membership: |
|

Genomic Map