Assay Details
Target Gene Details
Entrez Gene ID: | 80765 |
Gene Name: | StAR related lipid transfer domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.15:81312666-81324183 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD5 | NM_181900.2 | NP_871629.1 | ||
| NR_135013.1 | ||||
| XM_006720703.1 | XP_006720766.1 | |||
| XM_017022639.1 | XP_016878128.1 | |||
| AF480304.1 | AAL89654.1 | |||
| AK026352.1 | ||||
| AK097024.1 | ||||
| AK225479.1 | ||||
| AL137657.1 | CAB70862.2 | |||
| BC004365.1 | AAH04365.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048788 | Chr.15:81257750 - 81343926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map