Assay Details
Target Gene Details
Entrez Gene ID: | 100528031 |
Gene Name: | FKBP1A-SDCBP2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.20:1309909-1393172 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1A-SDCBP2 | NR_037661.1 | 10 | 1561 |
Target Gene Details
Entrez Gene ID: | 27111 |
Gene Name: | syndecan binding protein 2 |
Gene Aliases: |
SITAC, SITAC18, ST-2 |
Location: |
Chr.20:1309909-1329235 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDCBP2 | NM_001199784.1 | 9 | 1342 | NP_001186713.1 |
| NM_015685.5 | 5 | 1233 | NP_056500.2 | |
| NM_080489.4 | 9 | 1453 | NP_536737.3 | |
| AA514519.1 | 1 | 64 | ||
| AA907131.1 | 1 | 105 | ||
| AF131809.1 | 5 | 1199 | AAD20049.1 | |
| AF159228.1 | 9 | 1342 | AAF80369.1 | |
| BC002727.2 | 9 | 1392 | ||
| BM987490.1 | 1 | 143 | ||
| BU684857.1 | 1 | 73 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057097 | Chr.20:1050297 - 1333621 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map