Assay Details
Target Gene Details
Entrez Gene ID: | 654 |
Gene Name: | bone morphogenetic protein 6 |
Gene Aliases: |
VGR, VGR1 |
Location: |
Chr.6:7726778-7881728 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMP6 | NM_001718.4 | 1 | 399 | NP_001709.1 |
| AK300628.1 | 2 | 359 | ||
| BC096084.2 | AAH96084.1 | |||
| BC096085.2 | AAH96085.1 | |||
| BC099624.1 | AAH99624.1 | |||
| BC099625.1 | 2 | 131 | AAH99625.1 | |
| BC111763.1 | 2 | 131 | AAI11764.1 | |
| BC112352.1 | 2 | 131 | AAI12353.1 | |
| BC113018.1 | ||||
| BC113019.1 | ||||
| BC128031.1 | ||||
| M60315.1 | 1 | 380 | AAA36737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830582 | Chr.6:7706847 - 7889468 on Build GRCh38 | Loss |
|
| esv33520 | Chr.6:7726210 - 7729017 on Build GRCh38 | Gain |
|
| esv3890787 | Chr.6:6996928 - 7981438 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map