Assay Details
Target Gene Details
Entrez Gene ID: | 7389 |
Gene Name: | uroporphyrinogen decarboxylase |
Gene Aliases: |
PCT, UPD |
Location: |
Chr.1:45012133-45015669 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UROD | NM_000374.4 | NP_000365.3 | ||
| NR_036510.1 | ||||
| AF104421.1 | AAD04571.1 | |||
| AF104422.1 | AAD04572.1 | |||
| AF104423.1 | AAD04573.1 | |||
| AF104424.1 | AAD04574.1 | |||
| AF104425.1 | AAD04575.1 | |||
| AF104426.1 | AAD04576.1 | |||
| AF104427.1 | AAD04577.1 | |||
| AF104428.1 | AAD04578.1 | |||
| AF104429.1 | AAD04579.1 | |||
| AF104430.1 | AAD04580.1 | |||
| AF104431.1 | AAD04581.1 | |||
| AF104432.1 | AAD04582.1 | |||
| AF104433.1 | AAD04583.1 | |||
| AF104434.1 | AAD04584.1 | |||
| AF104435.1 | AAD04585.1 | |||
| AF104436.1 | AAD04586.1 | |||
| AF104437.1 | AAD04587.1 | |||
| AF104438.1 | AAD04588.1 | |||
| AF104439.1 | AAD04589.1 | |||
| AF104440.1 | AAD04590.1 |
More Information
Set Membership: |
|

Genomic Map