Assay Details
Target Gene Details
Entrez Gene ID: | 26574 |
Gene Name: | apoptosis antagonizing transcription factor |
Gene Aliases: |
BFR2, CHE-1, CHE1, DED |
Location: |
Chr.17:36948875-37056871 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AATF | NM_012138.3 | 3 | 871 | NP_036270.1 |
| XM_011524611.1 | 3 | 855 | XP_011522913.1 | |
| AF083208.1 | 3 | 788 | AAD52016.1 | |
| AJ249940.2 | 3 | 726 | CAB57451.2 | |
| AK026173.1 | 1 | 297 | ||
| AK057229.1 | 3 | 871 | ||
| BC000591.2 | 3 | 751 | AAH00591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833430 | Chr.17:36809156 - 37024342 on Build GRCh38 | Gain+Loss |
|
| esv3640480 | Chr.17:36761281 - 37124883 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map