Assay Details
Target Gene Details
Entrez Gene ID: | 84549 |
Gene Name: | MAK16 homolog |
Gene Aliases: |
MAK16L, RBM13 |
Location: |
Chr.8:33485167-33501260 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAK16 | NM_032509.3 | 10 | 1174 | NP_115898.2 |
| AF251062.1 | 10 | 1149 | AAK34952.1 | |
| AK027469.1 | 10 | 1159 | BAB55134.1 | |
| AK225476.1 | ||||
| AL049422.1 | ||||
| BC028230.1 | 10 | 1142 | AAH28230.1 | |
| BC039740.1 | 10 | 1155 | AAH39740.1 | |
| BC050528.1 | 10 | 1142 | AAH50528.1 |
Target Gene Details
Entrez Gene ID: | 80185 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
C8orf41, MRT39 |
Location: |
Chr.8:33498509-33513601 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTI2 | NM_001102401.2 | 8 | 2043 | NP_001095871.1 |
| NM_001265581.1 | 8 | 2165 | NP_001252510.1 | |
| NM_025115.3 | 7 | 2412 | NP_079391.2 | |
| AI890434.1 | 1 | 167 | ||
| AK308918.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76212197] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map